Assay Details
| Gene Symbol: CNOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192774.1 | NP_001179703.1 | 2 - 3 | 326 | 73 | |
| GenBank mRNA | BC119978.1 | - | 2 - 3 | 348 | 73 | |
More Information
Set Membership: |
|

CNOT8
Genomic Map