Assay Details
| Gene Symbol: SLCO4C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192846.1 | NP_001179775.1 | 8 - 9 | 1444 | 63 | |
| GenBank mRNA | AY885241.1 | - | 1 - 2 | 163 | 63 | |
More Information
Set Membership: |
|

SLCO4C1
Genomic Map