Assay Details
| Gene Symbol: RGS9BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166516.1 | NP_001159988.1 | 1 | 524 | 74 | |
| GenBank mRNA | AF480475.1 | - | 1 | 524 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGS9BP| Gene Symbol: RGS9BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166516.1 | NP_001159988.1 | 1 | 524 | 74 | |
| GenBank mRNA | AF480475.1 | - | 1 | 524 | 74 | |
Set Membership: |
|