Assay Details
| Gene Symbol: LIPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206597.1 | NP_001193526.1 | 2 - 3 | 262 | 62 | |
| GenBank mRNA | DV917166.1 | - | 3 - 4 | 244 | 62 | |
More Information
Set Membership: |
|

LIPM
Genomic Map