Assay Details
| Gene Symbol: SREK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206619.1 | NP_001193548.1 | 2 - 3 | 232 | 67 | |
| GenBank mRNA | BC118474.1 | - | 2 - 3 | 177 | 67 | |
More Information
Set Membership: |
|

SREK1IP1
Genomic Map