Assay Details
| Gene Symbol: ZNF74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206752.1 | NP_001193681.1 | 2 - 3 | 352 | 57 | |
| GenBank mRNA | EE387538.1 | - | 2 - 3 | 352 | 57 | |
More Information
Set Membership: |
|

ZNF74
Genomic Map