Assay Details
| Gene Symbol: ZNF74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206752.1 | NP_001193681.1 | 3 - 4 | 482 | 72 | |
| GenBank mRNA | DV929841.1 | - | 1 - 2 | 191 | 72 | |
| EE387538.1 | - | 3 - 4 | 482 | 72 | ||
More Information
Set Membership: |
|

ZNF74
Genomic Map