Assay Details
| Gene Symbol: PROCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166568.1 | NP_001160040.1 | 1 - 2 | 461 | 74 | |
| GenBank mRNA | BC151740.1 | - | 1 - 2 | 316 | 74 | |
More Information
Set Membership: |
|

PROCA1
Genomic Map