Assay Details
| Gene Symbol: DCAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167896.2 | NP_001161368.1 | 4 - 5 | 904 | 62 | |
| GenBank mRNA | BC105241.1 | - | 4 - 5 | 595 | 62 | |
| BC150062.1 | - | 4 - 5 | 903 | 62 | ||
More Information
Set Membership: |
|

DCAF12
Genomic Map