Assay Details
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172039.1 | NP_001165510.1 | 5 - 6 | 644 | 87 | |
| GenBank mRNA | BC150002.1 | - | 5 - 6 | 685 | 87 | |
| GU304600.1 | - | 5 - 6 | 644 | 87 | ||
More Information
Set Membership: |
|

RBMX
Genomic Map