Assay Details
| Gene Symbol: INS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185126.1 | NP_001172055.1 | 2 - 3 | 272 | 80 | |
| NM_173926.2 | NP_776351.2 | 2 - 3 | 256 | 80 | ||
| GenBank mRNA | BC142034.1 | - | 2 - 3 | 249 | 80 | |
| EH379095.1 | - | 2 - 3 | 270 | 80 | ||
| EV652245.1 | - | 2 - 3 | 302 | 80 | ||
| JX041514.1 | - | 1 - 2 | 119 | 80 | ||
More Information
Set Membership: |
|

INS
Genomic Map