Assay Details
| Gene Symbol: MTERF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191174.1 | NP_001178103.1 | 2 - 3 | 199 | 83 | |
| GenBank mRNA | BC151726.1 | - | 2 - 3 | 151 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151726.1, NM_001191174.1 transcripts.
Set Membership: |
|

MTERF2
Genomic Map