Assay Details
| Gene Symbol: PRMT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206873.1 | NP_001193802.1 | 7 - 8 | 867 | 64 | |
| GenBank mRNA | DV891499.1 | - | 4 - 5 | 378 | 64 | |
More Information
Set Membership: |
|

PRMT8
Genomic Map