Assay Details
| Gene Symbol: HRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242346.1 | NP_001229275.1 | 1 - 2 | 112 | 112 | |
| NM_001242347.1 | NP_001229276.1 | 1 - 2 | 112 | 112 | ||
| GenBank mRNA | CB419631.1 | - | 1 - 2 | 112 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001242346.1, NM_001242347.1 transcripts.
Set Membership: |
|

HRAS
Genomic Map