Assay Details
| Gene Symbol: HRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242346.1 | NP_001229275.1 | 2 - 3 | 269 | 72 | |
| NM_001242347.1 | NP_001229276.1 | 2 - 3 | 269 | 72 | ||
| GenBank mRNA | CB419631.1 | - | 2 - 3 | 269 | 72 | |
More Information
Set Membership: |
|

HRAS
Genomic Map