Assay Details
| Gene Symbol: HRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242346.1 | NP_001229275.1 | 3 - 4 | 448 | 65 | |
| NM_001242347.1 | NP_001229276.1 | 3 - 4 | 448 | 65 | ||
| GenBank mRNA | CB419631.1 | - | 3 - 4 | 448 | 65 | |
| DN532777.1 | - | 1 - 2 | 106 | 65 | ||
More Information
Set Membership: |
|

HRAS
Genomic Map