Assay Details
| Gene Symbol: OTUD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243311.1 | NP_001230240.1 | 11 - 12 | 932 | 64 | |
| GenBank mRNA | BC153856.1 | - | 8 - 9 | 1625 | 64 | |
| DV930774.1 | - | 1 - 2 | 88 | 64 | ||
More Information
Set Membership: |
|

OTUD4
Genomic Map