Assay Details
| Gene Symbol: NOX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191333.1 | NP_001178262.1 | 12 - 13 | 1576 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOX3| Gene Symbol: NOX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191333.1 | NP_001178262.1 | 12 - 13 | 1576 | 62 | |
Set Membership: |
|