Assay Details
| Gene Symbol: RAPSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191421.1 | NP_001178350.1 | 6 - 7 | 963 | 81 | |
| GenBank mRNA | DT893861.1 | - | 2 - 3 | 485 | 81 | |
| GH329633.1 | - | 4 - 5 | 392 | 81 | ||
More Information
Set Membership: |
|

RAPSN
Genomic Map