Assay Details
| Gene Symbol: MMP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191434.2 | NP_001178363.1 | 7 - 8 | 1369 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MMP15| Gene Symbol: MMP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191434.2 | NP_001178363.1 | 7 - 8 | 1369 | 68 | |
Set Membership: |
|