Assay Details
| Gene Symbol: LRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191469.1 | NP_001178398.1 | 11 - 12 | 1553 | 86 | |
| GenBank mRNA | EV724043.1 | - | 4 - 5 | 659 | 86 | |
More Information
Set Membership: |
|

LRWD1
Genomic Map