Assay Details
| Gene Symbol: FGF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245926.1 | NP_001232855.1 | 2 - 3 | 423 | 102 | |
| GenBank mRNA | EU693028.1 | - | 2 - 3 | 423 | 102 | |
More Information
Set Membership: |
|

FGF9
Genomic Map