Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183365.1 | NP_899209.1 | 3 - 4 | 373 | 95 | |
| GenBank mRNA | AF422181.1 | - | 3 - 4 | 373 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NCR1| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183365.1 | NP_899209.1 | 3 - 4 | 373 | 95 | |
| GenBank mRNA | AF422181.1 | - | 3 - 4 | 373 | 95 | |
Set Membership: |
|