Assay Details
| Gene Symbol: MAD2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191513.1 | NP_001178442.1 | 485 | 98 | ||
| GenBank mRNA | AB098952.1 | - | 99 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAD2L1| Gene Symbol: MAD2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191513.1 | NP_001178442.1 | 485 | 98 | ||
| GenBank mRNA | AB098952.1 | - | 99 | |||
Set Membership: |
|