Assay Details
| Gene Symbol: PTPN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192054.3 | NP_001178983.2 | 7 - 8 | 1165 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTPN9| Gene Symbol: PTPN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192054.3 | NP_001178983.2 | 7 - 8 | 1165 | 64 | |
Set Membership: |
|