Assay Details
| Gene Symbol: KATNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192103.1 | NP_001179032.1 | 10 - 11 | 1544 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KATNA1| Gene Symbol: KATNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192103.1 | NP_001179032.1 | 10 - 11 | 1544 | 65 | |
Set Membership: |
|