Assay Details
| Gene Symbol: SLC30A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192180.1 | NP_001179109.1 | 1 - 2 | 645 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC30A10| Gene Symbol: SLC30A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192180.1 | NP_001179109.1 | 1 - 2 | 645 | 56 | |
Set Membership: |
|