Assay Details
| Gene Symbol: PSMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192184.2 | NP_001179113.2 | 10 - 11 | 1311 | 70 | |
| GenBank mRNA | BC103063.1 | - | 10 - 11 | 1095 | 70 | |
| BT025458.1 | - | 10 - 11 | 1092 | 70 | ||
| EW699968.1 | - | 2 - 3 | 272 | 70 | ||
More Information
Set Membership: |
|

PSMC3
Genomic Map