Assay Details
| Gene Symbol: PGAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192191.1 | NP_001179120.1 | 1 - 2 | 232 | 53 | |
| GenBank mRNA | BC149319.1 | - | 1 - 2 | 225 | 53 | |
More Information
Set Membership: |
|

PGAM5
Genomic Map