Assay Details
| Gene Symbol: HEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192213.1 | NP_001179142.1 | 3 - 4 | 501 | 77 | |
| GenBank mRNA | BC114876.1 | - | 3 - 4 | 438 | 77 | |
More Information
Set Membership: |
|

HEBP2
Genomic Map