Assay Details
| Gene Symbol: PPP1R14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193070.2 | NP_001179999.1 | 2 - 3 | 478 | 94 | |
| GenBank mRNA | EE249911.1 | - | 1 - 2 | 78 | 94 | |
| EV633362.1 | - | 2 - 3 | 322 | 94 | ||
More Information
Set Membership: |
|

PPP1R14A
Genomic Map