Assay Details
| Gene Symbol: SDCCAG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193109.2 | NP_001180038.2 | 17 - 18 | 2267 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDCCAG8| Gene Symbol: SDCCAG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193109.2 | NP_001180038.2 | 17 - 18 | 2267 | 59 | |
Set Membership: |
|