Assay Details
| Gene Symbol: SLC18A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193171.1 | NP_001180100.1 | 11 - 12 | 1332 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC18A1| Gene Symbol: SLC18A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193171.1 | NP_001180100.1 | 11 - 12 | 1332 | 64 | |
Set Membership: |
|