Assay Details
| Gene Symbol: GFER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193188.1 | NP_001180117.1 | 1 - 2 | 265 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GFER| Gene Symbol: GFER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193188.1 | NP_001180117.1 | 1 - 2 | 265 | 80 | |
Set Membership: |
|