Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195029.1 | NP_001181958.1 | 1 - 2 | 32 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195029.1 transcripts.
Set Membership: |
|

LYRM9
Genomic Map