Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195029.1 | NP_001181958.1 | 3 - 4 | 265 | 103 | |
| GenBank mRNA | BC140576.1 | - | 2 - 3 | 339 | 103 | |
More Information
Set Membership: |
|

LYRM9
Genomic Map