Assay Details
| Gene Symbol: CRELD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046191.1 | NP_001039656.1 | 5 - 6 | 668 | 59 | |
| GenBank mRNA | BC112517.1 | - | 5 - 6 | 668 | 59 | |
More Information
Set Membership: |
|

CRELD2
Genomic Map