Assay Details
| Gene Symbol: GPR85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075150.3 | NP_001068618.2 | 1 - 2 | 116 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001075150.3 transcripts.
Set Membership: |
|

GPR85
Genomic Map