Assay Details
| Gene Symbol: RASSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076105.1 | NP_001069573.1 | 5 - 6 | 1084 | 64 | |
| GenBank mRNA | BC116069.1 | - | 5 - 6 | 1084 | 64 | |
More Information
Set Membership: |
|

RASSF7
Genomic Map