Assay Details
| Gene Symbol: NNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201324.1 | NP_001188253.1 | 1 - 2 | 164 | 76 | |
| GenBank mRNA | BC103128.1 | - | 1 - 2 | 165 | 76 | |
More Information
Set Membership: |
|

NNAT
Genomic Map