Assay Details
| Gene Symbol: SNRPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195027.1 | NP_001181956.1 | 3 - 4 | 334 | 102 | |
| GenBank mRNA | BC102192.1 | - | 3 - 4 | 288 | 102 | |
More Information
Set Membership: |
|

SNRPF
Genomic Map