Assay Details
| Gene Symbol: HAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075761.1 | NP_001069229.1 | 2 | 1421 | 111 | |
| GenBank mRNA | BC120210.1 | - | 2 | 1421 | 111 | |
More Information
Set Membership: |
|

HAND1
Genomic Map