Assay Details
| Gene Symbol: MIR2370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030980.1 | 1 | 42 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | filamin B | FLNB | Sense | Chr. 22 - 43132187 - 43272039 on NCBI Build ARS-UCD1.2 | ||
| Downstream | sarcolemma associated protein | SLMAP | 122.2kb | Sense | Chr. 22 - 43321043 - 43473168 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2370
Genomic Map