Assay Details
Gene Symbol: MIR2464 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031001.1 | 1 | 33 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | LysM domain containing 3 | LYSMD3 | -294.1kb | Anti-sense | Chr. 7 - 90083735 - 90097651 on NCBI Build ARS-UCD1.2 | |
Downstream | arrestin domain containing 3 | ARRDC3 | 447.7kb | Anti-sense | Chr. 7 - 90839580 - 90853625 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
