Assay Details
| Gene Symbol: MIR448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031055.1 | 1 | 53 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-1298 | MIR1298 | bta-mir-1298 | -83.4kb | Sense | Chr. X - 63075806 - 63075913 on NCBI Build ARS-UCD1.2 |
| Downstream |
RNA binding motif protein, X-linked 2-like |
LOC534630 | 153.4kb | Sense | Chr. X - 63312889 - 63323304 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR448
Genomic Map