Assay Details
| Gene Symbol: MIR2339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031111.1 | 1 | 38 | 56 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | elaC ribonuclease Z 2 | ELAC2 | Sense | Chr. 19 - 31361733 - 31378657 on NCBI Build ARS-UCD1.2 | ||
| Downstream | heparan sulfate-glucosamine 3-sulfotransferase 3A1 | HS3ST3A1 | 369.1kb | Sense | Chr. 19 - 31738492 - 31822369 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2339
Genomic Map