Assay Details
| Gene Symbol: MIR135B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030860.1 | 1 | 46 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LEM domain containing 1 | LEMD1 | -27.2kb | Sense | Chr. 16 - 3140265 - 3170241 on NCBI Build ARS-UCD1.2 | |
| Downstream | cyclin dependent kinase 18 | CDK18 | PCTK3 | 51.1kb | Anti-sense | Chr. 16 - 3248595 - 3275158 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR135B
Genomic Map