Assay Details
| Gene Symbol: SNHG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036663.1 | 2 - 3 | 150 | 116 | ||
| GenBank mRNA | EV667084.1 | - | 2 - 3 | 84 | 116 | |
| EW681979.1 | - | 2 - 3 | 150 | 116 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | regulator of chromosome condensation 1 | RCC1 | -13.0kb | Sense | Chr. 2 - 124880941 - 124903281 on NCBI Build ARS-UCD1.2 | |
| Downstream | phosphatase and actin regulator 4 | PHACTR4 | 5.6kb | Sense | Chr. 2 - 124922939 - 125032434 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

SNHG3
Genomic Map