Assay Details
| Gene Symbol: MGC134179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038137.1 | 1 | 725 | 100 | ||
| GenBank mRNA | BC111140.1 | - | 1 | 725 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | poly(A) binding protein cytoplasmic 5 | PABPC5 | -292.7kb | Sense | Chr. X - 39337380 - 39342039 on NCBI Build ARS-UCD1.2 | |
| Downstream | family with sequence similarity 133 member A | FAM133A | 2.5mb | Sense | Chr. X - 42120455 - 42171393 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MGC134179
Genomic Map