Assay Details
| Gene Symbol: SNHG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038147.1 | 1 - 2 | 703 | 111 | ||
| GenBank mRNA | EU848421.1 | - | 1 - 2 | 704 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TATA-box binding protein associated factor 12 | TAF12 | -16.5kb | Sense | Chr. 2 - 124810106 - 124834314 on NCBI Build ARS-UCD1.2 | |
| Downstream | tRNA selenocysteine 1 associated protein 1 | TRNAU1AP | TRSPAP1 | 2.2kb | Anti-sense | Chr. 2 - 124854126 - 124878715 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

SNHG12
Genomic Map